Universal-Fitting MicroFlex® Gel-Loading Pipette Tips, National Scientific Supply
Catalog # 53503-189
Supplier: Avantor
Specifications
- Volume:0.1 - 200 µl
- Cat. no.:53503-189
- Tip Style:Flat Tip
- Supplier No.:MN320F-LR
Specifications
About this item
MicroFlex® Gel-loading pipette tips are universal-fitting polypropylene pipette tips.
The tips are individually wrapped, Rnase and Dnase-free. Tips are not autoclavable.
Frequently Bought Together