Corning® Pasteur Pipettes
Disposable pasteur pipettes, Soda Lime Glass, 5.5 mm, 228.6 mm (9"), Non sterile
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
DISP PIPET 9IN PKG/250 9IN Catalog #KT883350-0009
Product KT883350-0009 is similar to 52947-802 but manufacturer and specifications may vary.
Product Details & Documents
Soda-lime or borosilicate glass.
- Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
- Features a constriction one inch below the top to facilitate plugging
- Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures.
Documents
Specifications
- Ext.Ø:5.5 mm
- Sterility:Non sterile
- Description:Disposable pasteur pipettes
- Length:228.6 mm (9")
- Material:Soda Lime Glass



