Order Entry
Canada
Orders LinkContactUsLinkComponent
Epredia™ Superfrost™ Plus Control Slides

Epredia™ Superfrost™ Plus Control Slides

Superfrost™ Plus, extended 1" white tab, 75×25 mm
Catalog # 48404-476
Supplier:   Erie Scientific
Supplier #:  ES-334PLUS-001
Epredia™ Superfrost™ Plus Control Slides
Catalog # 48404-476
Supplier:  Erie Scientific
Epredia™ Superfrost™ Plus Control Slides
Epredia™ Superfrost™ Plus Control Slides

Superfrost™ Plus, extended 1" white tab, 75×25 mm

Price
$127.41
New Customer Promotional Price
Log in for availability
Pack of 72 Case of 1440
Product Details & Documents

Ideal for staining procedures that require a control, these frosted slides feature spaces for both the patient sample and the control material

Slides may be used in histology, cytology, or microbiology applications. Slides are designed to ensure that tissue samples remain on the slide. A red or gold box identifies the control section. A red control box on the back surface identifies slides intended for use in automated instruments. Plus Gold control slides are recommended for hard-to-hold sections.

Specifications
  • Description:
    Superfrost™ Plus, extended 1" white tab
  • L×W:
    75×25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.