Orders LinkContactUsLinkComponent
VWR® Stopcocks
VWR® Stopcocks
15 mm (0.59''), 12.7 mm (0.5'')
Catalog # 46600-038
Supplier:   VWR International
VWR® Stopcocks
Catalog # 46600-038
Supplier:  VWR International
VWR® Stopcocks
VWR® Stopcocks
15 mm (0.59''), 12.7 mm (0.5'')
Price
$347.95
Log in for availability
Pack of 10 Case of 20
Product Details & Documents
These stopcocks are ideal for use with liquids and gases under low pressure (operating pressure 7.0psi) or low vacuum.
  • Ideal for low-pressure use
  • HDPE body
  • Polypropylene tap
  • Straight-bored stopcocks
The HDPE body and polypropylene tap provide a smooth turning action without the use of lubricants. Stopcocks are straight bored.
Documents
Specifications
  • Bore size:
    12.7 mm (0.5'')
  • Nominal O.D.:
    15 mm (0.59'')