Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Capsules, Hydrion®

pH Buffer Capsules, Hydrion®

±0.02, 7.00, Clear
Catalog # 34175-242
Supplier:   Micro Essential
Supplier #:  270-7.00
pH Buffer Capsules, Hydrion®
Catalog # 34175-242
Supplier:  Micro Essential
CAS Number:  
pH Buffer Capsules, Hydrion®
pH Buffer Capsules, Hydrion®

±0.02, 7.00, Clear

CAS Number:  
Price
$34.71
Log in for availability
Each Case of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

For the quick and convenient preparation of fresh solutions of known pH values

Capsule contents dissolve in 100mL of distilled water, giving a clear solution accurate to ±0.02 pH unit. The dry powder remains stable. Each vial contains 10 capsules.

Documents

Specifications

Specifications

  • Color:
    Clear
  • pH:
    7.00
  • pH accuracy:
    ±0.02

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.