Order Entry
Canada
Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

2 L, 8
Catalog # 29111-161
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  4060-2L
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Catalog # 29111-161
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

2 L, 8

Price
$107.52
New Customer Promotional Price
Log in for availability
Case of 1
Product Details & Documents

Manufactured from borosilicate glass.

  • High thermal shock resistance
  • Provide contamination free results
  • High chemical durability

Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).

Specifications
  • Capacity:
    2 L
  • Stopper no.:
    8
  • Cat. No.:
    29111-161
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact