Orders LinkContactUsLinkComponent
PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning
PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning
Extractor Cup
Catalog # 27614-434
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  3915-ECO
PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning
Catalog # 27614-434
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Accelerated Modular One-Step™ Extraction Apparatus, Corning
Extractor Cup
Price
482,00 $
Log in for availability
Case of 1
Product Details & Documents
This revolutionary modular extractor-concentrator performs extraction, concentration and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.
Removable Snyder column sidearm allows easy cleaning. Extraction and concentration steps can be performed outside a hood, although the use of a ventilation system is recommended above or near the units in case of a drop in condenser water temperature or loss of condenser cold water supply.
Certifications: Meets EPA test requirements outlined by SW-846 method 3520 and Waste Water method 625 allowed in CLP statement of work OLMO2.0 for semi-volatiles and pesticides.
Ordering information: This apparatus requires a PYREX® concentrator tube (27614-428) and PYREX® Allihn condenser (see 27613-022 series), snyder sidearm, extractor cup and body, O-rings, connectors, membrane and stopcock assembly and membrane support should be ordered separately..
Documents
Specifications
  • Description:
    Extractor Cup
  • Cat. No.:
    27614-434
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact