Split Stoppers, Labconco®
Split Stopper, 20 mm Corkage
Catalog # 26676-253
Supplier: Labconco
Supplier #: 7576210
Jump to:
Product Details & Documents
Split Stoppers
Package of 100. Rubber stopper, when crimped with an aluminum seal, tightly seals Serum Bottle for lyophilization applications. For use with FreeZone Freeze Dry Systems. Stoppers are for serum bottles with 20 mm corkage or 13 mm corkage, as indicated.
Documents
Labconco FreeZone® Freeze Dryers and Accessories
Product Literature
Specifications
- Description:Split Stopper, 20 mm Corkage
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



