Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffers, YSI

pH Buffers, YSI

pH Buffer, ±0.01, Box of 6 Pints, 475 ml, 4, red
Catalog # 23202-821
Supplier:   YSI
Supplier #:  3821
pH Buffers, YSI
Catalog # 23202-821
Supplier:  YSI
CAS Number:  
pH Buffers, YSI

pH Buffer, ±0.01, Box of 6 Pints, 475 ml, 4, red

CAS Number:  
Price
$42.52
Log in for availability
Each Pack of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

pH buffers for pH probe calibration.

Each buffer case contains 6 individual pints (plastic). Buffer cases are available with a single buffer type or with an assortment of buffers (2 pints of pH 4, 7, and 10 buffers).

Specifications

Specifications

  • Color:
    red
  • Description:
    pH Buffer
  • Size:
    475 ml
  • Packed:
    Box of 6 Pints
  • pH:
    4
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.