Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Push Brooms, Perfex

Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3"), PP, Yellow, 61 cm (24")
  22940-294
 :   Perfex
 :  2624Y
Push Brooms, Perfex
  22940-294
 :  Perfex
Push Brooms, Perfex
Push Brooms, Perfex

Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3"), PP, Yellow, 61 cm (24")

 
75,06 $
   
Each
 

Synthetic brushes are designed to exceed the rigorous demands of today’s controlled environments

Polypropylene fibers are fused onto a high impact-resistant polymer block to eliminate premature fiber fallout that could contaminate collection areas. Lite-N-Tite® handle/broom head connection system locks handle securely in place. Brush and handle are nonconductive and will not absorb bacteria, liquids, or odors. Both are unaffected by water, petroleum products, detergents, sanitizers, and solvents. Durable, lightweight, sanitary adjustable handle, made of high-quality polyester and composite materials, reduces strain and transfers maximum energy with minimum exertion.

 : Brush heads are available in two sizes and five colors for light/medium and heavy sweeping tasks. A special extra-heavy model is also available. Brush heads and handles sold separately.

 
  • Bristle color:
    Yellow
  • Bristle material:
    PP
  • Description:
    Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3")
  • Head width:
    61 cm (24")
Accessories

 

  •  
    Description
    Handle Material
    Handle Color
     
     
  • 53200-100
    Adjustable Handle with Long Connector
    Polymer
    White
       
    Each - 141,82 $

 

  •  
    Description
    Handle Material
    Handle Color
     
     
  • 22940-104
    Fixed-Length Handle
    Polymer
    White
       
    Each - 77,26 $

 

  •  
    Description
    Dimensions
    Handle Length
     
     
  • 11310-906
    Hardwood Handle, 60", Natural
    2.5 cm (1") Diameter
    152 cm (60")
       
    Each - 19,76 $