Short Handle Scrub Brushes, Perfex
PP, Green, Plastic, 9 5/8", 1¾", 3"
22940-276
: Perfex
Supplier #: 3020G
This short handle scrub brush contains stiff polypropylene fiber made for aggressive scrubbing of baked-on deposits.
- Conforms to irregular surfaces and will not collapse under powerful scrubbing
- Durable fiber maintains shape and is visually coded to isolate areas of use
- Brush block and fiber will not absorb bacteria, liquids or odors
- Will not rot, chip or decay
- Unaffected by water, grease, petroleum products, detergents, sanitizer, and solvents
Brush fiber is heat-fused into impact-resistant polymer brush block, eliminating premature fiber fallout and areas for contaminants to collect.
- Bristle color:Green
- Bristle material:PP
- Handle material:Plastic
- Total length:9 5/8"
- Width:3"
- Trim length:1¾"