Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Short Handle Scrub Brushes, Perfex

PP, White, Plastic, 9 5/8", 1¾", 3"
  22940-202
 :   Perfex
 :  3020W
Short Handle Scrub Brushes, Perfex
  22940-202
 :  Perfex
Short Handle Scrub Brushes, Perfex
Short Handle Scrub Brushes, Perfex

PP, White, Plastic, 9 5/8", 1¾", 3"

 
$66.79
   
Each Case of 10
 

This short handle scrub brush contains stiff polypropylene fiber made for aggressive scrubbing of baked-on deposits.

  • Conforms to irregular surfaces and will not collapse under powerful scrubbing
  • Durable fiber maintains shape and is visually coded to isolate areas of use
  • Brush block and fiber will not absorb bacteria, liquids or odors
  • Will not rot, chip or decay
  • Unaffected by water, grease, petroleum products, detergents, sanitizer, and solvents

Brush fiber is heat-fused into impact-resistant polymer brush block, eliminating premature fiber fallout and areas for contaminants to collect.

 
  • Bristle color:
    White
  • Bristle material:
    PP
  • Handle material:
    Plastic
  • Total length:
    9 5/8"
  • Width:
    3"
  • Trim length:
    1¾"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.