Order Entry
Canada
Orders LinkContactUsLinkComponent
Limited Volume Inserts, WHEATON®

Limited Volume Inserts, WHEATON®

Insert for vial with step, 0.25 ml, Glass, No
Catalog # 16364-998
Supplier #:  225258
Limited Volume Inserts, WHEATON®
Catalog # 16364-998
Limited Volume Inserts, WHEATON®
Limited Volume Inserts, WHEATON®

Insert for vial with step, 0.25 ml, Glass, No

Price
$302.40
New Customer Promotional Price
Log in for availability
Case of 100
Product Details & Documents

For E-Z Vials (12×32 mm) with wide opening.

  • Inserts fit nicely into the vials
  • Available with or without writing patch
  • Glass and polypropylene limited volume inserts

Purchase vials and snap caps separately or together in convenience packs.

Packaging: Packaged in shrink-wrapped trays of 100 for convenience and cleanliness.

Specifications
  • Cap included:
    No
  • Capacity:
    0.25 ml
  • Description:
    Insert for vial with step
  • Material:
    Glass
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.