Orders LinkContactUsLinkComponent
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
Sample Cup for Technicon® and Other Autoanalyzers, 4 ml (0.14 oz.)
Catalog # 15070-292
Supplier #:  K2508-VWR
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
Catalog # 15070-292
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
Sample Cup for Technicon® and Other Autoanalyzers, 4 ml (0.14 oz.)
Price
$1,894.35
Log in for availability
Case of 5,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These transparent polystyrene sample cups are resistant to most dilute aqueous solutions.
  • Compatible with most autoanalyzers
  • Wide base prevents spillage
Cups are available for use with Technicon®, Abbott®, Hitachi®, and most other autoanalyzers
Autoanalyzer sample cups are not autoclavable.
The wide-base design prevents tipping and spilling. The conical bottom is standard and recommended for microsampling.
Ordering information: The 2.0 mL cup (15070-290) also fits the Hyland laser-beam analyzer. Push-on polyethylene cap (15070-089) fits cups 15070-290, -293, and -294.
Specifications
  • Capacity:
    4 ml (0.14 oz.)
  • Description:
    Sample Cup for Technicon® and Other Autoanalyzers
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Polyethylene Cap
    Polyethylene Cap
    15070-089
    Log in for availability
    Case of 1,000 - $143.40
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)