Order Entry
Canada
Orders LinkContactUsLinkComponent
PYREX® Bell Jars, Corning

PYREX® Bell Jars, Corning

457 mm (18"), 457 mm (18")
Catalog # 14183-306
Supplier:   Corning
Supplier #:  6886-49L
PYREX® Bell Jars, Corning
Catalog # 14183-306
Supplier:  Corning
PYREX® Bell Jars, Corning
PYREX® Bell Jars, Corning

457 mm (18"), 457 mm (18")

Price
$6,190.00
Log in for availability
Case of 1
Product Details & Documents

These specially blown, sturdy, round-bottom jars have a fine ground finish at the open end. Flat, ground surface has a contact area equal to the wall thickness at the open end.

Caution: Do not use with heat, pressure or vacuum applications.

Documents

Specifications
  • Ext.Ø:
    457 mm (18")
  • Height:
    457 mm (18")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.