Orion™ Basic Electrodes, Thermo Scientific
Low Maintenance Gel Combination Electrode, with built-in ATC, Epoxy Body
Catalog # 14004-290
Supplier: Thermo Fisher Scientific
Supplier #: 9147BN
Jump to:
Product Details & Documents
Perform basic pH measurements quickly and reliably with Orion™ Basic Electrodes, for use with Orion™ Russell meters.
- pH range of 0 to 14
Specifications
- Description:Low Maintenance Gel Combination Electrode, with built-in ATC
- Material:Epoxy Body
- Cat. No.:14004-290
- Operating temperature range:0 to 80°C