$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
SLEEVE TEF. FLUOKEM PK3 19/38
Catalog # 13196-014
Supplier:
Bel-Art Products, a Part of SP
Expand image
SLEEVE TEF. FLUOKEM PK3 19/38
Catalog # 13196-014
Supplier:
Bel-Art Products, a Part of SP
Supplier Number:
F20414-1938
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH