Low Density Flat Poly Liners, Associated Bag
Yellow, 33 gal. cans (liner circumference 66"), 33×40 ", 4 mil.
Catalog # 11215-918
Supplier: Associated Bag
Supplier #: 71-6-35Y
Jump to:
Product Details & Documents
Sort and identify different types of waste.
Strong, bottom-sealed liners are free of heavy metals, making them safe for landfill or incineration.
Specifications
- Color:Yellow
- Thickness:4 mil.
- W×L:33×40 "
- Fits:33 gal. cans (liner circumference 66")
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



