Orders LinkContactUsLinkComponent
ClipperMop™ Cleanroom Mops
ClipperMop™ Cleanroom mop with head assembly and telescoping handle, 18 x 10 cm (7 x 4")
  TWTX7102
 :   Texwipe
 :  TX7102
ClipperMop™ Cleanroom Mops
  TWTX7102
 :  Texwipe
ClipperMop™ Cleanroom Mops
ClipperMop™ Cleanroom Mops
ClipperMop™ Cleanroom mop with head assembly and telescoping handle, 18 x 10 cm (7 x 4")
521,90 $
Each
The mop system may be used with wet, dry or sterile wipers, and is compatible with protocols for ISO Class 3 to 8 cleanrooms, sterile and aseptic areas
  • Cleanroom compatible mopping system
  • Patent-pending thermoplastic mop with clips
  • Quick and easy wiper change
  • Lightweight, easy to handle
  • Less expensive than competitors cleanroom mops
The small size conveniently cleans hard-to-reach areas, walls, ceilings, floors and equipment exteriors. The mop system may be sterilized by gamma irradiation, chemicals, vaporized hydrogen peroxide (VHP) or ethylene oxide. Widely used in pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries.
The patent-pending mophead utilizes non-abrasive polyethylene clips to securely fasten one or multiple wipers to the head. The ClipperMop wiper attachment method does not release particles, fibers and dirt.
 : The ClipperMop™ cleanroom mop includes the Celcon® thermoplastic mop head with clips, a flexible polyester pad for better wiping efficiency and an adjustable handle.
  • Description:
    ClipperMop™ Cleanroom mop with head assembly and telescoping handle
  • Dimensions:
    18 x 10 cm (7 x 4")
Accessories
  •  
    Description
  • TWTX7102C
    ClipperMop™ tether clip replacement kit, 4 polyethylene tether clips and 2 nylon screws per bag
    Case of 20 - 322,04 $
  •  
    Description
  • TWTX7111B
    ClipperMop™ replacement pads, foam pads (101,6×177,8) and white fasteners for use with TX7103
    Bag of 10 - 13,58 $