Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Wright-Giemsa stain

Glass bottle, 1 L
Catalog # RC938032
Health
Toxic
Irritant
Flammable
Supplier:   Ricca Chemical
 :  R9380000-1C
Wright-Giemsa stain
Catalog # RC938032
Supplier:  Ricca Chemical
CAS Number:  
Wright-Giemsa stain
Wright-Giemsa stain

Glass bottle, 1 L

CAS Number:  
Price
$316.50
Log in for availability
Each (1l)
 
Product Details & Documents

Wright-Giemsa Stain Mixture

Synonyms:  Wright-Giemsa Solution
  • MDL Number:MFCD00132593
  • Boiling Pt:~ 64.5 °C
  • Density:0.8484 g/cm³
  • Flash Pt:~ 12 °C (MeOH)
  • Storage Temperature:Ambient
  • UN:1993
  • ADR:3,II
Specifications

Specification Test Results

  • Absorbance (traceable to NIST SRM 930):
    Conforms to control
  • Appearance:
    Dark blue, Methanol odor

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L

Specifications