Ammonia solution 2%
Poly bottle, 1 L
Supplier: Ricca Chemical
Supplier #: R0631000-1A
Jump to:
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Boiling Pt:100 °C
- Melting Pt:0.0 °C
- Density:0.9978 g/cm³
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Sulfuric Acid/Methyl Red):1.90 - 2.10 % (v/v)
Specifications
- Pack type:Poly bottle
- Size:1 L
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



