Order Entry
Canada
Orders LinkContactUsLinkComponent
Acetate Buffer solutions, pH 4.00, Ricca Chemical Company

Acetate Buffer solutions, pH 4.00, Ricca Chemical Company

±0.1, Acetate, 25 °C, Poly bottle, 1 gal., 4.00, Colorless
Catalog # RC501
Supplier:   Ricca Chemical
Supplier #:  R0050000-4A
Acetate Buffer solutions, pH 4.00, Ricca Chemical Company
Catalog # RC501
Supplier:  Ricca Chemical
CAS Number:  
Acetate Buffer solutions, pH 4.00, Ricca Chemical Company
Acetate Buffer solutions, pH 4.00, Ricca Chemical Company

±0.1, Acetate, 25 °C, Poly bottle, 1 gal., 4.00, Colorless

CAS Number:  
Price
$465.15
Log in for availability
Each (1US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Acetate buffer, pH 4.0, for residual chlorine analysis

Standardized at 25°C against chemicals whose certification is traceable to NIST.

Documents

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Color:
    Colorless
  • Size:
    1 gal.
  • pH buffer type:
    Acetate
  • Reference temperature:
    25 °C
  • pH:
    4.00
  • pH accuracy:
    ±0.1

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.