Order Entry
Canada
Orders LinkContactUsLinkComponent
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company

Buffers, Reference Standards, Color-Coded, Ricca Chemical Company

±0.01, Carbonate, 25 °C, Cubetainer®, 10 L, 10.00, Blue
Catalog # RC160125
Supplier:   Ricca Chemical
Supplier #:  R1601000-10F
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Catalog # RC160125
Supplier:  Ricca Chemical
CAS Number:  
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company

±0.01, Carbonate, 25 °C, Cubetainer®, 10 L, 10.00, Blue

CAS Number:  
Price
$254.63
Log in for availability
Each (320oz)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Certified traceable to NIST Standard Reference Materials
  • Lot numbers and expiration dates
Specifications

Specifications

  • Pack type:
    Cubetainer®
  • Color:
    Blue
  • Size:
    10 L
  • pH buffer type:
    Carbonate
  • Reference temperature:
    25 °C
  • pH:
    10.00
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.