Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
±0.01, Phthalate, 25 °C, Poly bottle, 1 L, 4.00, Red
Supplier: Ricca Chemical
Supplier #: R1501000-1A
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Catalog # RC150132
Supplier: Ricca Chemical
CAS Number:
Jump to:
Product Details & Documents
- Certified traceable to NIST Standard Reference Materials
- Lot numbers and expiration dates
Documents
Certificates
Search by Lot NumberRC1501-16
Images 360
Ricca Chemical Company Buffer Reference Standards Flyer
Product Literature
Specifications
Specifications
- Pack type:Poly bottle
- Color:Red
- Size:1 L
- pH buffer type:Phthalate
- Reference temperature:25 °C
- pH:4.00
- pH accuracy:±0.01
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



