Order Entry
Canada
Orders LinkContactUsLinkComponent
D-(-)-Lactic acid

D-(-)-Lactic acid

1 g
Catalog # Q-1870.1000BA
Supplier:   Bachem Americas
Supplier #:  4003254.1000
D-(-)-Lactic acid
Catalog # Q-1870.1000BA
Supplier:  Bachem Americas
CAS Number:  10326-41-7
D-(-)-Lactic acid

1 g

CAS Number:  10326-41-7
Price
$986.61
Log in for availability
Each (1g)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequence: D-Lactic acid

Useful chiral synthons, building blocks for depsipeptides.

Synonym(s): (R)-2-Hydroxy-propionic acid#H-D-Lac-OH

Synonyms:  (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
  • CAS Number:10326-41-7
  • Formula:C₃H₆O₃
  • MDL Number:MFCD00068313
  • Molecular Weight:90.08
  • Storage Temperature:Refrigerator
  • EINECS:233-713-2
Specifications

Specifications

  • Size:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.