D-(-)-Lactic acid
250 mg
Catalog # Q-1870.0250BA
Supplier: Bachem Americas
Supplier #: 4003254.0250
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Sequence: D-Lactic acid
Useful chiral synthons, building blocks for depsipeptides.
Synonym(s): (R)-2-Hydroxy-propionic acid#H-D-Lac-OH
Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
- CAS Number:10326-41-7
- Formula:C₃H₆O₃
- MDL Number:MFCD00068313
- Molecular Weight:90.08
- Storage Temperature:Refrigerator
- EINECS:233-713-2
Specifications
Specifications
- Size:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



