Pepstatin A
25 mg
Catalog # N-1125.0025BA
Supplier: Bachem Americas
: 4031136.0025
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Pepstatin A (isovaleryl-pepstatin), a low molecular weight inhibitor of acid proteases, e.g. pepsin, renin, cathepsin D, and plasmepsin II. For the Plasmodium falciparum aspartyl protease plasmepsin II, Hill et al determined an IC₅₀ of 0.1 nM.
Specifications
Specifications
- Size:25 mg
- Cat. No.:N-1125.0025BA
- Molecular formula:C₃₄H₆₃N₅O₉
- Protein/peptide name:Pepstatin A
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00060740
- Sequence:Isovaleryl-Val-Val-Sta-Ala-Sta-OH
- Molecular weight:685.90
- CAS No.:26305-03-3



