Orders LinkContactUsLinkComponent
Micro-Vials, DWK Life Sciences
Micro-Vials, DWK Life Sciences
5.00 ml, No
Catalog # KT746100-0005
Supplier #:  746100-0005
Micro-Vials, DWK Life Sciences
Catalog # KT746100-0005
Micro-Vials, DWK Life Sciences
Micro-Vials, DWK Life Sciences
5.00 ml, No
Price
$61.15
New Customer Promotional Price
Log in for availability
Case of 1
Product Details & Documents
Glassware manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Standard taper 14/10 connections
  • Ideal for use up to 500 °C
Specifications
  • Cap included:
    No
  • Capacity:
    5.00 ml