Orders LinkContactUsLinkComponent
KIMBLE® TLC Rectangular Developing Tanks, DWK Life Sciences
KIMBLE® TLC Rectangular Developing Tanks, DWK Life Sciences
Developing tank with standard lid, 27×7×25 cm (105/8×23/4×97/8")
Catalog # KT416180-0000
Supplier #:  416180-0000
KIMBLE® TLC Rectangular Developing Tanks, DWK Life Sciences
Catalog # KT416180-0000
KIMBLE® TLC Rectangular Developing Tanks, DWK Life Sciences
KIMBLE® TLC Rectangular Developing Tanks, DWK Life Sciences
Developing tank with standard lid, 27×7×25 cm (105/8×23/4×97/8")
Price
$454.75
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Sturdy molded glass developing tanks feature clear sides to allow unobstructed visual inspection of TLC plates up to 20x20cm (77/8x77/8") in size
Tanks are available with a standard, nonslip, or latch style lid. The nonslip lid has a knob for easy lifting, and PTFE retainers inside prevent the lid from sliding off the tank. The latch lid holds lid firmly to the tank, creating a stable environment for developing TLC plates. The latch lid also has a knob for easy lifting and PTFE retainers inside.
Nonslip and latch tank lids are available with holes drilled for #0 rubber stoppers to facilitate the addition of reagents with minimal disruption to the environment of developing plates.
Edges are beveled to eliminate sharp edges. Tanks also have a ground bottom and top for a level surface. A raised edge along the inside bottom allows the simultaneous development of two 20x20cm (77/8x77/8") or four 5x20cm (2x77/8") plates.
Weight: 5.4 kg (11.9 lbs.)
Ordering information: Rack needs to be ordered seperately.
Documents
Specifications
  • Description:
    Developing tank with standard lid
  • W×D×H:
    27×7×25 cm (105/8×23/4×97/8")
Accessories
Product Information
  • Catalog #
    No. of Places
    Availability
    Price
  • TLC aluminum rack
    TLC aluminum rack
    KT416175-0000
    6
    Log in for availability
    Each - $603.50
Product Information
  • Catalog #
    Availability
    Price
  • TLC Tank Latch Lid
    TLC Tank Latch Lid
    KT416191-0000
    Log in for availability
    Each - $215.05
Product Information
  • Catalog #
    Availability
    Price
  • TLC Developing Tank for use with Latch Lid
    TLC Developing Tank for use with Latch Lid
    KT416192-0000
    Log in for availability
    Each - $604.80
Product Information
  • Catalog #
    Availability
    Price
  • TLC Tank Non-Slip Lid
    TLC Tank Non-Slip Lid
    KT416186-0000
    Log in for availability
    Each - $230.40
Product Information
  • Catalog #
    Availability
    Price
  • TLC Standard Lid
    TLC Standard Lid
    KT416181-0000
    Log in for availability
    Case of 1 - $85.85
Product Information
  • Catalog #
    Availability
    Price
  • TLC Developing Tank without Lid
    TLC Developing Tank without Lid
    KT416182-0000
    Log in for availability
    Each - $422.45