Mouse;Rat ACTH (1-39) Trifluoroacetate
H-4998.1000BA
:
- CAS No.:77465-10-2
- Species:RatMouse
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00167431
- Protein synonyms:Adrenocorticotropic hormone
- Protein/peptide name:ACTH (1-39) Trifluoroacetate
- Molecular weight:4582.23
- Sequence:H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Val-Ala-Glu-Asn-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OH
- Molecular formula:C₂₁₀H₃₁₅N₅₇O₅₇S
- Cat. no.:H-4998.1000BA
- Supplier No.:H-4998.1000
Please see also α-MSH (corresponds to acetyl-ACTH (1-13) amide, H-1075), α-MSH (free acid) (acetyl-ACTH (1-13), H-1070), and (Des-acetyl)-α-MSH (H-4390). For reviews on ACTH and related peptides see: D.de Wied: Pituitary Adrenal System Hormones and Behaviour. Symposium on Developments in Endocrinology (1976) / Anon.: ACTH and Related Peptides: Structure, Regulation, and Action. Ann. N.Y. Acad. Sci. 297, 1 (1977) / A.V.Schally: Aspects of Hypothalamic Regulation of the Pituitary Gland. Science 202, 18 (1978) / R.Schwyzer: Studies on Polypeptide Receptors. A Critical View on the Mechanism of ACTH Action. Bull. Schweiz. Acad. Med. Wiss. 34, 263 (1978).
Frequently Bought Together