Order Entry
Canada
Orders LinkContactUsLinkComponent
trans-4-Hydroxy-L(-)-proline

trans-4-Hydroxy-L(-)-proline

100 g
Catalog # F-1655.0100BA
Supplier:   Bachem Americas
Supplier #:  4002357.0100
trans-4-Hydroxy-L(-)-proline
Catalog # F-1655.0100BA
Supplier:  Bachem Americas
CAS Number:  51-35-4
trans-4-Hydroxy-L(-)-proline

100 g

CAS Number:  51-35-4
Price
489,17 $
Log in for availability
Each (100g)
Minimum order quantity is 1
Product Details & Documents

Sequence: H-Hyp-OH

Hyp denotes L-trans-hydroxyproline.

Synonym(s): (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid

Synonyms:  L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
  • CAS Number:51-35-4
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064320
  • Molecular Weight:131.13 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-091-9
Specifications

Specifications

  • Size:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.