Order Entry
Canada
Orders LinkContactUsLinkComponent
H-Cys(StBu)-OH

H-Cys(StBu)-OH

5 g
Catalog # E-2710.0005BA
Supplier:   Bachem Americas
Supplier #:  4030538.0005
H-Cys(StBu)-OH
Catalog # E-2710.0005BA
Supplier:  Bachem Americas
CAS Number:  30044-51-0
H-Cys(StBu)-OH

5 g

CAS Number:  30044-51-0
Price
$187.24
Log in for availability
Each (5g)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequence: H-Cys(StBu)-OH

Synonyms:  S-tert-Butylmercapto-L-cysteine
  • CAS Number:30044-51-0
  • Formula:C₇H₁₅NO₂S₂
  • MDL Number:MFCD00002609
  • Molecular Weight:209.322 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Size:
    5 g

Specifications