Order Entry
Canada
Orders LinkContactUsLinkComponent
Comparators for CHEMets® Test Kits

Comparators for CHEMets® Test Kits

Low Range Ammonia Comparator, 0 - 1 ppm, 0, 0.1, 0.2, 0.3, 0.4, 0.6 0.8, 1.0 ppm
Catalog # CXC-1501
Supplier:   CHEMetrics
Supplier #:  C-1501
Comparators for CHEMets® Test Kits
Catalog # CXC-1501
Supplier:  CHEMetrics
CAS Number:  
Comparators for CHEMets® Test Kits
Comparators for CHEMets® Test Kits

Low Range Ammonia Comparator, 0 - 1 ppm, 0, 0.1, 0.2, 0.3, 0.4, 0.6 0.8, 1.0 ppm

CAS Number:  
Price
$47.83
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: Store in the dark at room temperature.

Documents

Specifications

Specifications

  • Description:
    Low Range Ammonia Comparator
  • Increments:
    0, 0.1, 0.2, 0.3, 0.4, 0.6 0.8, 1.0 ppm
  • Range:
    0 - 1 ppm

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.