Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TLC PLATE,SI-C18 PK-25 20X20CM
CAJT7012-4
:
AVANTOR PERFORMANCE MATERIAL LLC
TLC PLATE,SI-C18 PK-25 20X20CM
CAJT7012-4
:
AVANTOR PERFORMANCE MATERIAL LLC
:
7012-04
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International