Urea ≥99% USP
Fiber drum, Leverpack®, 110 lbs.
Jump to:
Discontinued
Product Details & Documents
GMP manufactured.
- GMP manufactured product
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:13,09935
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:132.7 °C
- Density:1.34 g/cm³ (20 °C)
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
Specifications
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Assay (NH₂CONH₂):98.0 - 102.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Alcohol Insoluble Matter:≤0.04 %
- USP - Organic impurities: Urea related compound A:≤0.1 %
- USP - Organic impurities: Any individual unspecified impurity:≤0.1 %
- USP - Organic impurities: Total impurities:≤2.0 %
Specifications
- Pack type:Fiber drum, Leverpack®
- Size:110 lbs.
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



