Orders LinkContactUsLinkComponent
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Needle with Pre-Attached Holder, 3.175 cm (11/4"), 21 G
Catalog # CABD368650
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Catalog # CABD368650
Supplier:  
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Needle with Pre-Attached Holder, 3.175 cm (11/4"), 21 G
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
NEEDLE BD ECLIPSE 21G PK48 Catalog #CABD368607L
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These sterile, safety-engineered, multi-sample blood collection needles offer a simple, effective way to collect blood while reducing the possibility of needlestick injuries
They feature a patented safety shield that allows for one-handed activation to cover the needle completely upon withdrawal from the vein, and confirm proper activation with an audible click. The safety shield is already aligned with the bevel of the needle, so no extra assembly or adjustment is required by the user. The intuitive design of the advanced protection technology means there is no need to change the standard, one-handed venipuncture technique when using these needles. Needles with pre-attached holders help facilities remain single-use holder compliant.
Specifications
  • Description:
    Needle with Pre-Attached Holder
  • Needle length:
    3.175 cm (11/4")
  • Needle gauge:
    21 G
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.