Formic acid ≥85%
Product Details & Documents
Purity: 85% CAS No.: 64-18-6 Molecular Formula: CH2O2 Formula Weight: 46.03 Flash Point: 50°(156°F) Refractive Index: 1.3710 Melting Point: 7-9° Boiling Point: 100-101° Beilstein Reference: 1209246 Merck Reference: 14,4241 UN No.: 1779 MDL No.: MFCD00003297
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03
- Merck Index:14,4241
- Boiling Pt:100…101 °C
- Melting Pt:7…9 °C
- Density:1.220
- Flash Pt:50 °C (156 °F)
- Storage Temperature:Ambient
- UN:1779
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
Documents
Specifications
Specifications
- Pack type:Bottle
- Size:2.5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



