Orders LinkContactUsLinkComponent
1,6-Hexanedithiol 97%
Bottle, 5 g
  CAAAAL06686-06
 :  L06686-06
1,6-Hexanedithiol 97%
  CAAAAL06686-06
 :  1191-43-1
1,6-Hexanedithiol 97%
Bottle, 5 g
 :  1191-43-1
$48.26
Each (5g)
Purity: 97% CAS No.: 1191-43-1 Molecular Formula: HS(CH2)6SH Formula Weight: 150.30 Flash Point: 90°(194°F) Refractive Index: 1.5130 Melting Point: -21° Boiling Point: 242-243° Beilstein Reference: 1732507 UN No.: 2810 MDL No.: MFCD00004910
 :  1,6-Dimercaptohexane
  •  :1191-43-1
  •  :C₆H₁₄S₂
  •  :MFCD00004910
  •  :150.31 g/mol
  •  :242…243 °C
  •  :–21 °C
  •  :0.990
  •  :90 °C(194 °F)
  •  :Ambient
  •  :214-735-1
  •  :3334
  •  :9,
  • Pack type:
    Bottle
  • Size:
    5 g
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.