Order Entry
Canada
Orders LinkContactUsLinkComponent
Phenol red

Phenol red

Bottle, 250 g
Catalog # CAAAAB21710-30
Irritant
Supplier #:  B21710-30
Phenol red
Catalog # CAAAAB21710-30
CAS Number:  143-74-8
Phenol red
Phenol red

Bottle, 250 g

CAS Number:  143-74-8
Price
$584.79
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Odor: Odorless Boiling Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Merck Index:14,7247
  • Boiling Pt:>300 °C
  • Melting Pt:> 300 °C
  • Storage Temperature:Ambient
  • EINECS:205-609-7

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.