Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium hydroxide ≥85%, pellets

Potassium hydroxide ≥85%, pellets

Bottle, 2.5 kg
Catalog # CAAAAA18854-0E
Corrosive
Irritant
Supplier #:  A18854-0E
Potassium hydroxide ≥85%, pellets
Catalog # CAAAAA18854-0E
CAS Number:  1310-58-3
Potassium hydroxide ≥85%, pellets

Bottle, 2.5 kg

CAS Number:  1310-58-3
Price
$176.93
Log in for availability
Each (2,500kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 85% CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Odor: Odorless pH: 13,5 Melting Point: 360° Boiling Point: 1320-1324° Merck Reference: 14,7640 UN No.: 1813 MDL No.: MFCD00003553

Synonyms:  Potash caustic, Potassium hydrate, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Merck Index:14,7640
  • Boiling Pt:1320…1324 °C
  • Melting Pt:360 °C
  • Density:2.044
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1813
  • ADR:8,II
  • REACH:01-2119487136-33

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    2.5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.