Order Entry
Canada
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99+%

L(+)-Ascorbic acid 99+%

Bottle, 500 g
Catalog # CAAAAA15613-36
Supplier #:  A15613-36
L(+)-Ascorbic acid 99+%
Catalog # CAAAAA15613-36
CAS Number:  50-81-7
L(+)-Ascorbic acid 99+%
L(+)-Ascorbic acid 99+%

Bottle, 500 g

CAS Number:  50-81-7
Price
$134.99
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 190-193° dec. Beilstein Reference: 84272 Merck Reference: 14,830 Optical Rotation: +21° (c=10 in water) MDL No.: MFCD00064328

Synonyms:  Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:14,830
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…193 °C (dec.)
  • Density:1.65
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.