Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Bismuth(III) carbonate basic

Bottle, 250 g
Catalog # CAAAAA12483-30
 :  A12483-30
Bismuth(III) carbonate basic
Catalog # CAAAAA12483-30
CAS Number:  5892-10-4
Bismuth(III) carbonate basic

Bottle, 250 g

CAS Number:  5892-10-4
Price
$160.89
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 Formula Weight: 509.97 Merck Reference: 14,1282 MDL No.: MFCD00010877

Synonyms:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS Number:5892-10-4
  • Formula:(BiO)₂CO₃
  • MDL Number:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Merck Index:14,1282
  • Density:6.86
  • Storage Temperature:Ambient
  • EINECS:227-567-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.