1-(2-Pyridylazo)-2-naphthol (PAN) solution 0,1% indicator
Dropping bottle, self-contained, 50 ml
Catalog # CA97026-624
Supplier: Hach
: 2122426
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0,1% indicator
Catalog # CA97026-624
Supplier: Hach
CAS Number: 85-85-8
Jump to:
Product Details & Documents
For determination of low level manganese by the PAN method
Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.
Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol
- CAS Number:85-85-8
- Formula:C₁₅H₁₁N₃O
- MDL Number:MFCD00004071
- Molecular Weight:249.27 g/mol
- Storage Temperature:Ambient
- UN:3082
- ADR:9,III
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Method:8149
- Method Name:PAN
- Parameter:Manganese
- Platform :Solution
- Range:0.006 - 0.700 mg/L Mn
Specifications
- Pack type:Dropping bottle, self-contained
- Size:50 ml



