Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Rectangular Semi-Demountable Cell Windows, PerkinElmer

Calcium Fluoride Cell Windows, 41 mm, 4 mm, 23 mm
Catalog # CA97013-766
Supplier:   PerkinElmer
 :  L1271193
Rectangular Semi-Demountable Cell Windows, PerkinElmer
Catalog # CA97013-766
Supplier:  PerkinElmer
Rectangular Semi-Demountable Cell Windows, PerkinElmer
Rectangular Semi-Demountable Cell Windows, PerkinElmer

Calcium Fluoride Cell Windows, 41 mm, 4 mm, 23 mm

Price
$1,323.23
Log in for availability
Pack of 2
Product Details & Documents

Rectangular windows make the demountable cell becomes a semi-demountable cell. It can be used either as a demountable cell or as a sealed cell for high-viscosity liquids or mull samples.

  • Available in a variety of window materials
  • Windows come in pairs

Combining two different requirements into one cell allows a myriad of different sampling applications.

 : Semi-demountable Cell can be ordered with the universal demountable cell, windows and spacers.

Specifications
  • Width:
    23 mm
  • Description:
    Calcium Fluoride Cell Windows
  • Height:
    41 mm
  • Thickness:
    4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.