Order Entry
Canada
Orders LinkContactUsLinkComponent
NIST Traceable pH Buffers with Temperature Table

NIST Traceable pH Buffers with Temperature Table

Bottle, 500 ml, 4.00
Catalog # CA97007-784
Supplier:   Mettler Toledo
Supplier #:  63056121
NIST Traceable pH Buffers with Temperature Table
Catalog # CA97007-784
Supplier:  Mettler Toledo
CAS Number:  
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table

Bottle, 500 ml, 4.00

CAS Number:  
Price
$36.30
Log in for availability
Each (500ml)
Product Details & Documents

NIST-traceable buffer for pH electrode calibration and verification.

  • Temperature table on bottle

Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml
  • pH:
    4.00

Specifications