Orders LinkContactUsLinkComponent
Lab Worktables, Mobile, Metro
Lab Worktables, Mobile, Metro
Mobile worktables with solid HD shelf, Gray, Phenolic resin, Stainless steel, 35⅞", 35¾×30", 92 lbs.
Catalog # CA97002-806
Supplier:   Metropolitan Wire
Supplier #:  LTSM36PG
Lab Worktables, Mobile, Metro
Catalog # CA97002-806
Supplier:  Metropolitan Wire
Lab Worktables, Mobile, Metro
Lab Worktables, Mobile, Metro
Mobile worktables with solid HD shelf, Gray, Phenolic resin, Stainless steel, 35⅞", 35¾×30", 92 lbs.
Price
$3,376.75
Log in for availability
Each
Product Details & Documents
Stainless steel lab worktables provides corrosion-proof performance.
  • Tables feature smooth, easy-to-clean surfaces
  • Tops are made of Type 304 stainless steel or Trespa® phenolic resin
  • Phenolic resin tops are compression molded of organic fiber-reinforced phenolic core with an integrally cured thermoset resin surface
  • Phenolic resin tops are ideal for moderate to high chemical use and have excellent abrasion resistance and impact resistance
Tables are load rated at 244 kg/m2 (50lbs./ft.2) up to a maximum of 272 kg (600lbs.), assuming evenly distributed load and caster specifications that meet requirements. Tops with backsplash feature a 10.2cm (4") high backsplash.
Ordering information: Casters for mobile worktables must be ordered separately; for casters and other accessories, see 97000-762 series.
Specifications
  • Base/leg material:
    Stainless steel
  • Worksurface material:
    Phenolic resin
  • Description:
    Mobile worktables with solid HD shelf
  • Height:
    35⅞"
  • W×D:
    35¾×30"
  • Shipping weight:
    92 lbs.
  • Worksurface color/finish:
    Gray
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Stainless steel drawer
    Stainless steel drawer
    CA97002-830
    Log in for availability
    Each - $999.58
Product Information
  • Catalog #
    Color
    Material
    Availability
    Price
  • Swivel stem caster, high modulus rubber donut tread
    Swivel stem caster, high modulus rubber donut tread
    CA19766-816
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; Advantage high modulus gray thermoplastic rubber donut tread; Polyolefin hub
    Log in for availability
    Each - $137.64
Product Information
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel/brake stem caster, 300 lbs. load capacity, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    Swivel/brake stem caster, 300 lbs. load capacity, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    CA97005-858
    5"
    2.3 lbs.
    Log in for availability
    Case of 8 - $657.53
Product Information
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel/brake stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    Swivel/brake stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    CA97002-900
    5"
    2 lbs.
    Log in for availability
    Each - $308.66
Product Information
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    Swivel stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    CA97002-902
    5"
    2.1 lbs.
    Log in for availability
    Each - $576.71
Product Information
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Swivel stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    Swivel stem caster, antimicrobial polyurethane, gray, nylon horn with nickel plated cold rolled steel stem and stainless steel C-ring; antimicrobial polyurethane flat tread; polyethylene hub
    CA97002-898
    5"
    2 lbs.
    Log in for availability
    Each - $287.29
Product Information
  • Catalog #
    Diameter
    Shipping Weight
    Availability
    Price
  • Rigid stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    Rigid stem caster, polyurethane, silver/gray, stainless steel horn; polyurethane flat tread; polyethylene hub
    CA97005-860
    5"
    2 lbs.
    Log in for availability
    Each - $599.83
Product Information
  • Catalog #
    Color
    Material
    Availability
    Price
  • Swivel/brake stem caster, high modulus rubber donut tread
    Swivel/brake stem caster, high modulus rubber donut tread
    CA22221-208
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; Advantage high modulus gray thermoplastic rubber donut tread; polyolefin hub
    Log in for availability
    Each - $159.49
Product Information
  • Catalog #
    Availability
    Price
  • Stainless steel keyboard tray
    Stainless steel keyboard tray
    CA97002-828
    Log in for availability
    Each - $980.89