Swingsette™ M517 Tissue Processing/Embedding Cassettes, Simport
Disposable plastic cassettes hold tissue specimens during processing, embedding, and storage
A special hinge holds the base and lid together and allows cassettes to be opened and closed as often as necessary. The lid can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
Most convenient for cassette labeling instruments, as the lids are packaged separately in the case. They are molded from a special, high-density acetal polymer and are completely resistant to the chemical action of histological solvents. Cassettes keep specimens safely submerged in liquid while the efficient flow-through slots maximize fluid exchange and ensure proper drainage.
: Each case contains two dispenser boxes of 500 cassettes and one dispenser box of 1000 lids.
- Color:Yellow
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.