Orders LinkContactUsLinkComponent
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Gray
Catalog # CA97000-460
Supplier:   Simport Scientific
 :  M515-9
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Catalog # CA97000-460
Supplier:  Simport Scientific
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Swingsette™ M515 Tissue Processing/Embedding Cassettes, Simport
Gray
Price
$496.53
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Disposable plastic cassettes hold tissue specimens during processing, embedding, and storage
A special hinge holds the base and cover together and allows cassettes to be opened and closed as often as necessary. The cover can be removed and reinserted easily without the danger of specimen loss. Anterior surface is slanted at a 45° angle for use with leading labeling instruments. Three frosted writing surfaces. Cassette colors are non-cytotoxic and non-metallic.
They are molded from a special, high-density acetal polymer and are completely resistant to the chemical action of histological solvents. Cassettes keep specimens safely submerged in liquid while the efficient flow-through slots maximize fluid exchange and ensure proper drainage.
 : Each case contains three dispenser boxes of 500 cassettes with the lids closed.
Specifications
  • Color:
    Gray
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)