Orders LinkContactUsLinkComponent
Slimsette™ M511 Tissue Processing/Embedding Cassettes, Simport
Slimsette™ M511 Tissue Processing/Embedding Cassettes, Simport
Orange
Catalog # CA97000-436
Supplier:   Simport Scientific
Supplier #:  M511-11
Slimsette™ M511 Tissue Processing/Embedding Cassettes, Simport
Catalog # CA97000-436
Supplier:  Simport Scientific
Slimsette™ M511 Tissue Processing/Embedding Cassettes, Simport
Slimsette™ M511 Tissue Processing/Embedding Cassettes, Simport
Orange
Price
$603.24
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These compact cassettes feature a unique recessed cover that allows more cassettes to be stacked in automatic labeling machines and in storage cabinets
Cassettes are molded from acetal polymer and are completely resistant to the chemical action of histological solvents. Four 13mm square compartments have openings in the cover and base for maximizing fluid exchange and ensuring proper drainage. The one-piece integral lid eliminates the need for separate steel lids and can be opened and closed as often as necessary. Lids relock securely without danger of specimen loss. Cassettes feature large labeling areas for easy identification. The anterior writing area is slanted at a 45° angle for suitability with certain types of cassette labeling instruments. Cassette colors are non-cytotoxic and non-metallic.
Ordering information: Each case contains three dispenser boxes of 500 cassettes with the lids closed.
Specifications
  • Color:
    Orange
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.