Order Entry
Canada
Orders LinkContactUsLinkComponent
Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific

Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific

Tissue Cassettes, Blue
Catalog # CA95030-294
Supplier:   Simport Scientific
Supplier #:  M517-6SL
Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Catalog # CA95030-294
Supplier:  Simport Scientific
Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific

Tissue Cassettes, Blue

Price
$300.84
Log in for availability
Case of 750
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid

The cassettes are totally resistant to the chemical action of histological solvents. The flow-through openings allow for maximum fluid exchange and ensure proper drainage. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle. All cassettes colors are non-cytotoxic and non-metallic. Dimensions: 42.2Lx28.5Wx6Hmm (121/32x11/8x1/4").

Ordering information: Each case contains 10 sleeves of 75 bases and 10 bags of 75 lids. Cassette bases are packaged in transparent sleeves for quick loading into automatic labeling instruments.

Documents

Specifications
  • Color:
    Blue
  • Description:
    Tissue Cassettes

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.